COM448685372 Combi-Blocks, Inc.

2-Fluoro-5-(thiophen-3-yl)benzoic acid

  • MDL number MFCD18318887
  • Cas Number : 1261993-00-3
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8747

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8747
Availability details for
Quantity
  • Availability
  • Add to Cart