COM448685373 Combi-Blocks, Inc.

5-Fluoro-3-(thiophen-3-yl)benzoic acid

  • MDL number MFCD18318888
  • Cas Number : 1261925-85-2
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8748

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8748
Availability details for
Quantity
  • Availability
  • Add to Cart