COM448685374 Combi-Blocks, Inc.

5-Methoxy-3-(thiophen-3-yl)benzoic acid

  • MDL number MFCD18318893
  • Cas Number : 1261969-21-4
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8753
Availability details for
Quantity
  • Availability
  • Add to Cart