COM448685379 Combi-Blocks, Inc.

3-Fluoro-4-(2-methylphenyl)benzoic acid

  • MDL number MFCD16509409
  • Cas Number : 1284813-06-4
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8776

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8776
Availability details for
Quantity
  • Availability
  • Add to Cart