COM448685383 Combi-Blocks, Inc.

5-Methoxy-3-(2-methylphenyl)benzoic acid

  • MDL number MFCD18318918
  • Cas Number : 1262004-87-4
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8784

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8784
Availability details for
Quantity
  • Availability
  • Add to Cart