COM448685384 Combi-Blocks, Inc.

2-Chloro-4-(2-methylphenyl)benzoic acid

  • MDL number MFCD18318920
  • Cas Number : 1238634-51-9
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8787

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8787
Availability details for
Quantity
  • Availability
  • Add to Cart