COM448685385 Combi-Blocks, Inc.

4-Chloro-3-(2-methylphenyl)benzoic acid

  • MDL number MFCD18318924
  • Cas Number : 1261969-09-8
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8791

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8791
Availability details for
Quantity
  • Availability
  • Add to Cart