COM448685386 Combi-Blocks, Inc.

3-(2-Methylphenyl)-5-nitrobenzoic acid

  • MDL number MFCD18318926
  • Cas Number : 1261952-79-7
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8793

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8793
Availability details for
Quantity
  • Availability
  • Add to Cart