COM448685395 Combi-Blocks, Inc.

3-(3-Methylphenyl)-5-trifluoromethylbenzoic acid

  • MDL number MFCD18318949
  • Cas Number : 1261953-10-9
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8826

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8826
Availability details for
Quantity
  • Availability
  • Add to Cart