COM448685396 Combi-Blocks, Inc.

4-(4-Methylphenyl)-3-methylbenzoic acid

  • MDL number MFCD18318951
  • Cas Number : 1261942-99-7
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8828

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8828
Availability details for
Quantity
  • Availability
  • Add to Cart