COM448685399 Combi-Blocks, Inc.

3-Fluoro-4-(4-methylphenyl)benzoic acid

  • MDL number MFCD16509402
  • Cas Number : 1261953-35-8
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8838

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8838
Availability details for
Quantity
  • Availability
  • Add to Cart