COM448685401 Combi-Blocks, Inc.

2-Fluoro-5-(4-methylphenyl)benzoic acid

  • MDL number MFCD12859181
  • Cas Number : 1183641-87-3
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8840

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8840
Availability details for
Quantity
  • Availability
  • Add to Cart