COM448685402 Combi-Blocks, Inc.

5-Fluoro-3-(4-methylphenyl)benzoic acid

  • MDL number MFCD18318959
  • Cas Number : 1261978-89-5
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8841

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8841
Availability details for
Quantity
  • Availability
  • Add to Cart