COM448685410 Combi-Blocks, Inc.

4-(4-Hydroxyphenyl)-3-methoxybenzoic acid

  • MDL number MFCD18319064
  • Cas Number : 1261901-65-8
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8967

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8967
Availability details for
Quantity
  • Availability
  • Add to Cart