COM448685411 Combi-Blocks, Inc.

4-(4-Hydroxyphenyl)-2-methoxybenzoic acid

  • MDL number MFCD18319068
  • Cas Number : 1261892-57-2
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8971
Availability details for
Quantity
  • Availability
  • Add to Cart