COM448685412 Combi-Blocks, Inc.

3-(2-Fluorophenyl)-4-fluorobenzoic acid

  • MDL number MFCD14699218
  • Cas Number : 1214340-35-8
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8992

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-8992
Availability details for
Quantity
  • Availability
  • Add to Cart