COM448685421 Combi-Blocks, Inc.

5-(3-Fluorophenyl)-2-fluorobenzoic acid

  • MDL number MFCD12859261
  • Cas Number : 1178958-75-2
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9026

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9026
Availability details for
Quantity
  • Availability
  • Add to Cart