COM448685423 Combi-Blocks, Inc.

3-(3-Fluorophenyl)-5-methoxybenzoic acid

  • MDL number MFCD14699836
  • Cas Number : 1214324-49-8
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9032

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9032
Availability details for
Quantity
  • Availability
  • Add to Cart