COM448685425 Combi-Blocks, Inc.

4-(3-Fluorophenyl)-2-nitrobenzoic acid

  • MDL number MFCD14700015
  • Cas Number : 887244-17-9
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9040

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9040
Availability details for
Quantity
  • Availability
  • Add to Cart