COM448685427 Combi-Blocks, Inc.

4-(4-Fluorophenyl)-2-hydroxybenzoic acid

  • MDL number MFCD14701332
  • Cas Number : 67531-84-4
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9051

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9051
Availability details for
Quantity
  • Availability
  • Add to Cart