COM448685432 Combi-Blocks, Inc.

3-(4-Fluorophenyl)-5-fluorobenzoic acid

  • MDL number MFCD14699222
  • Cas Number : 1214330-80-9
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9058

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9058
Availability details for
Quantity
  • Availability
  • Add to Cart