COM448685436 Combi-Blocks, Inc.

3-(3-Cyanophenyl)-5-hydroxybenzoic acid

  • MDL number MFCD18086682
  • Cas Number : 1258637-09-0
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9112

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9112
Availability details for
Quantity
  • Availability
  • Add to Cart