COM448685437 Combi-Blocks, Inc.

3-(3-Cyanophenyl)-4-fluorobenzoic acid

  • MDL number MFCD18319125
  • Cas Number : 1261977-67-6
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9116

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9116
Availability details for
Quantity
  • Availability
  • Add to Cart