COM448685438 Combi-Blocks, Inc.

4-(3-Cyanophenyl)-3-fluorobenzoic acid

  • MDL number MFCD16509408
  • Cas Number : 1261960-89-7
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9117

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9117
Availability details for
Quantity
  • Availability
  • Add to Cart