COM448685439 Combi-Blocks, Inc.

5-(3-Cyanophenyl)-2-fluorobenzoic acid

  • MDL number MFCD12859211
  • Cas Number : 1183926-58-0
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9119

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9119
Availability details for
Quantity
  • Availability
  • Add to Cart