COM448685441 Combi-Blocks, Inc.

3-(3-Cyanophenyl)-2-methoxybenzoic acid

  • MDL number MFCD18319129
  • Cas Number : 1261928-47-5
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9123

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9123
Availability details for
Quantity
  • Availability
  • Add to Cart