COM448685447 Combi-Blocks, Inc.

2-Fluoro-4-(2-formylphenyl)benzoic acid

  • MDL number MFCD18319182
  • Cas Number : 1261945-86-1
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9180

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9180
Availability details for
Quantity
  • Availability
  • Add to Cart