COM448685448 Combi-Blocks, Inc.

2-Chloro-5-(2-formylphenyl)benzoic acid

  • MDL number MFCD18319191
  • Cas Number : 1261958-60-4
  • Molecular Weight : 260.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9189

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9189
Availability details for
Quantity
  • Availability
  • Add to Cart