COM448685449 Combi-Blocks, Inc.

2-Chloro-4-(2-formylphenyl)benzoic acid

  • MDL number MFCD18319192
  • Cas Number : 1261970-85-7
  • Molecular Weight : 260.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9190

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9190
Availability details for
Quantity
  • Availability
  • Add to Cart