COM448685458 Combi-Blocks, Inc.

3-(4-Formylphenyl)-5-trifluoromethylbenzoic acid

  • MDL number MFCD18319262
  • Cas Number : 1261960-97-7
  • Molecular Weight : 294.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9260

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9260
Availability details for
Quantity
  • Availability
  • Add to Cart