COM448685461 Combi-Blocks, Inc.

4-(3,5-Dimethylphenyl)-3-methylbenzoic acid

  • MDL number MFCD18319361
  • Cas Number : 1261971-14-5
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9386

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9386
Availability details for
Quantity
  • Availability
  • Add to Cart