COM448685462 Combi-Blocks, Inc.

3-(3,5-Dimethylphenyl)-2-methylbenzoic acid

  • MDL number MFCD18319362
  • Cas Number : 1261892-78-7
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9387
Availability details for
Quantity
  • Availability
  • Add to Cart