COM448685467 Combi-Blocks, Inc.

3-(2-Methoxyphenyl)-2-methylbenzoic acid

  • MDL number MFCD18319476
  • Cas Number : 1261902-11-7
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9511

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9511
Availability details for
Quantity
  • Availability
  • Add to Cart