COM448685475 Combi-Blocks, Inc.

2-Fluoro-4-(3-methoxyphenyl)benzoic acid

  • MDL number MFCD12859254
  • Cas Number : 1178418-69-3
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9552
Availability details for
Quantity
  • Availability
  • Add to Cart