COM448685481 Combi-Blocks, Inc.

2-(4-Methoxyphenyl)-6-methylbenzoic acid

  • MDL number MFCD18319515
  • Cas Number : 1261902-83-3
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9571
Availability details for
Quantity
  • Availability
  • Add to Cart