COM448685482 Combi-Blocks, Inc.

4-(4-Methoxyphenyl)-3-methylbenzoic acid

  • MDL number MFCD18319516
  • Cas Number : 108478-59-7
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9572
Availability details for
Quantity
  • Availability
  • Add to Cart