COM448685486 Combi-Blocks, Inc.

5-Fluoro-3-(4-methoxyphenyl)benzoic acid

  • MDL number MFCD18319521
  • Cas Number : 1261903-69-8
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9585

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9585
Availability details for
Quantity
  • Availability
  • Add to Cart