COM448685489 Combi-Blocks, Inc.

4-(4-Methoxyphenyl)-2-nitrobenzoic acid

  • MDL number MFCD18319531
  • Cas Number : 669002-29-3
  • Molecular Weight : 274.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9598
Availability details for
Quantity
  • Availability
  • Add to Cart