COM448685490 Combi-Blocks, Inc.

3-(4-Methoxyphenyl)-5-trifluoromethylbenzoic acid

  • MDL number MFCD18319534
  • Cas Number : 1261902-93-5
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9601

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9601
Availability details for
Quantity
  • Availability
  • Add to Cart