COM448685497 Combi-Blocks, Inc.

2-Fluoro-4-(3-fluoro-4-methylphenyl)benzoic acid

  • MDL number MFCD18319633
  • Cas Number : 1261961-48-1
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9707

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9707
Availability details for
Quantity
  • Availability
  • Add to Cart