COM448685500 Combi-Blocks, Inc.

3-(3-Fluoro-4-methylphenyl)-5-nitrobenzoic acid

  • MDL number MFCD18319649
  • Cas Number : 1261903-43-8
  • Molecular Weight : 276.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9723

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9723
Availability details for
Quantity
  • Availability
  • Add to Cart