COM448685509 Combi-Blocks, Inc.

4-(4-Fluoro-2-hydroxyphenyl)-3-hydroxybenzoic acid

  • MDL number MFCD18319759
  • Cas Number : 1261956-28-8
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9855

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9855
Availability details for
Quantity
  • Availability
  • Add to Cart