COM448685510 Combi-Blocks, Inc.

4-(2-Chlorophenyl)-2-fluorobenzoic acid

  • MDL number MFCD12862948
  • Cas Number : 505082-99-5
  • Molecular Weight : 250.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9986

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9986
Availability details for
Quantity
  • Availability
  • Add to Cart