COM448685511 Combi-Blocks, Inc.

5-(2-Chlorophenyl)-2-fluorobenzoic acid

  • MDL number MFCD12862947
  • Cas Number : 1184837-79-3
  • Molecular Weight : 250.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9987

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9987
Availability details for
Quantity
  • Availability
  • Add to Cart