COM448685513 Combi-Blocks, Inc.

2-Chloro-4-(2-chlorophenyl)benzoic acid

  • MDL number MFCD18319888
  • Cas Number : 1237117-14-4
  • Molecular Weight : 267.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9996

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-9996
Availability details for
Quantity
  • Availability
  • Add to Cart