COM448685518 Combi-Blocks, Inc.

3-(3-Chlorophenyl)-5-fluorobenzoic acid

  • MDL number MFCD18319905
  • Cas Number : 1261929-09-2
  • Molecular Weight : 250.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0020

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0020
Availability details for
Quantity
  • Availability
  • Add to Cart