COM448685519 Combi-Blocks, Inc.

3-(3-Chlorophenyl)-5-methoxybenzoic acid

  • MDL number MFCD18319909
  • Cas Number : 1261920-84-6
  • Molecular Weight : 262.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0025

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0025
Availability details for
Quantity
  • Availability
  • Add to Cart