COM448685523 Combi-Blocks, Inc.

3-(4-Chlorophenyl)-2-methylbenzoic acid

  • MDL number MFCD18319922
  • Cas Number : 1261930-53-3
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0039

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0039
Availability details for
Quantity
  • Availability
  • Add to Cart