COM448685525 Combi-Blocks, Inc.

3-(4-Chlorophenyl)-4-fluorobenzoic acid

  • MDL number MFCD11933569
  • Cas Number : 1181639-60-0
  • Molecular Weight : 250.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0047

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0047
Availability details for
Quantity
  • Availability
  • Add to Cart