COM448685529 Combi-Blocks, Inc.

2-Chloro-4-(4-chlorophenyl)benzoic acid

  • MDL number MFCD18319931
  • Cas Number : 334018-55-2
  • Molecular Weight : 267.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0059

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0059
Availability details for
Quantity
  • Availability
  • Add to Cart